Methyl-2-amino-5-brombenzoat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-amino-5-bromobenzoate

OR4689-100G OR4689-25G
APOSOR4689-100GEA 78.3 EUR
APOSOR4689-100G APOSOR4689-25G
Methyl-2-amino-5-brombenzoat 98%
Methyl-2-amino-5-brombenzoat
Formel: C₈H₈BrNO₂
Molecular Weight: 230,06 g/mol
Schmelzpunkt: 72…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00144761
CAS-Nummer: 52727-57-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR