Methyl-2-brom-5-nitrobenzoat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Brom-5-nitrobenzoesäuremethylester
OR4695-100G
APOSOR4695-100GEA
112
EUR
APOSOR4695-100G
Methyl-2-brom-5-nitrobenzoat 98%
Methyl-2-brom-5-nitrobenzoat
Formel:
C₈H₆BrNO₄ Molecular Weight: 260,04 g/mol Schmelzpunkt: 79…81 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010867 CAS-Nummer: 6942-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...