5-Bromo-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2,3-dihydro-7-azaindole

OR470006-1G OR470006-5G
APOSOR470006-1GEA 63.8 EUR
APOSOR470006-1G APOSOR470006-5G
5-Bromo-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 98%
5-Bromo-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine
Formel: C₇H₇BrN₂
Molecular Weight: 199.0479 g/mol
Schmelzpunkt: 186…191 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659750
CAS-Nummer: 115170-40-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR