4-(Methansulfonylamino)benzonitril

Lieferant: Apollo Scientific

Synonyme: 4-(Methylsulphonylamino)benzonitrile

OR470012-25G OR470012-5G
APOSOR470012-25GEA 522 EUR
APOSOR470012-25G APOSOR470012-5G
4-(Methansulfonylamino)benzonitril
4-(Methansulfonylamino)benzonitril
Formel: C₈H₈N₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00204724
CAS-Nummer: 36268-67-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR