4-(4-Amino-2-chlorophenyl)-2-piperazinone
Lieferant: Apollo Scientific
OR470029-1G
OR470029-250MG
OR470029-5G
APOSOR470029-1GEA
713
EUR
APOSOR470029-1G
APOSOR470029-250MG
APOSOR470029-5G
4-(4-Amino-2-chlorophenyl)-2-piperazinone
4-(4-Amino-2-chlorophenyl)-2-piperazinone
Formel:
C₁₀H₁₂ClN₃O Molecular Weight: 225.68 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09043886 CAS-Nummer: 926250-84-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...