1-Pyrimidin-2-ylpiperidin-4-amine

Lieferant: Apollo Scientific

Synonyme: 4-Amino-1-(2-pyrimidinyl)piperidine

OR470032-1G OR470032-5G OR470032-250MG
APOSOR470032-1GEA 478 EUR
APOSOR470032-1G APOSOR470032-5G APOSOR470032-250MG
1-Pyrimidin-2-ylpiperidin-4-amine
1-Pyrimidin-2-ylpiperidin-4-amine
Formel: C₉H₁₄N₄
Molecular Weight: 178.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06616818
CAS-Nummer: 412355-81-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR