4-(5-Cyclopropyl-1,2,4-oxadiazol-3-yl)benzoic acid
Lieferant: Apollo Scientific
OR470038-5G
OR470038-1G
APOSOR470038-5GEA
1010
EUR
APOSOR470038-5G
APOSOR470038-1G
4-(5-Cyclopropyl-1,2,4-oxadiazol-3-yl)benzoic acid
4-(5-Cyclopropyl-1,2,4-oxadiazol-3-yl)benzoic acid
Formel:
C₁₂H₁₀N₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08691678 CAS-Nummer: 915920-27-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...