2-[2-(Methylsulphanyl)phenoxy]acetic acid

Lieferant: Apollo Scientific

Synonyme: [2-(Methylthio)phenoxy]acetic acid

OR470099-1G OR470099-5G
APOSOR470099-1GEA 215 EUR
APOSOR470099-1G APOSOR470099-5G
2-[2-(Methylsulphanyl)phenoxy]acetic acid
2-[2-(Methylsulphanyl)phenoxy]acetic acid
Formel: C₉H₁₀O₃S
Molecular Weight: 198,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09972274
CAS-Nummer: 3395-40-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR