[4-(Methylthio)phenoxy]acetic acid
Lieferant: Apollo Scientific
OR470100-1G
OR470100-5G
APOSOR470100-1GEA
194
EUR
APOSOR470100-1G
APOSOR470100-5G
[4-(Methylthio)phenoxy]acetic acid
[4-(Methylthio)phenoxy]acetic acid
Formel:
C₉H₁₀O₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00233485 CAS-Nummer: 15267-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...