3-(2-Cyanphenoxy)propionsäure

Lieferant: Apollo Scientific

OR470103-1G OR470103-5G
APOSOR470103-1GEA 306 EUR
APOSOR470103-1G APOSOR470103-5G
3-(2-Cyanphenoxy)propionsäure
3-(2-Cyanphenoxy)propionsäure
Formel: C₁₀H₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09803402
CAS-Nummer: 916198-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR