3-(4-Carbamoylphenoxy)propionsäure

Lieferant: Apollo Scientific

OR470105-1G OR470105-5G
APOSOR470105-1GEA 306 EUR
APOSOR470105-1G APOSOR470105-5G
3-(4-Carbamoylphenoxy)propionsäure
3-(4-Carbamoylphenoxy)propionsäure
Formel: C₁₀H₁₁NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD09948835
CAS-Nummer: 111140-92-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR