3-(2-Carbamoylphenoxy)propionsäure
Lieferant: Apollo Scientific
OR470106-1G
OR470106-5G
APOSOR470106-1GEA
306
EUR
APOSOR470106-1G
APOSOR470106-5G
3-(2-Carbamoylphenoxy)propionsäure
3-(2-Carbamoylphenoxy)propionsäure
Formel:
C₁₀H₁₁NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD02364319 CAS-Nummer: 103204-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...