Pentane-2,3,4-trione 3-oxime

Lieferant: Apollo Scientific

Synonyme: 3-Isonitroso-2,4-pentanedione

OR470145-5G OR470145-1G
APOSOR470145-5GEA 578 EUR
APOSOR470145-5G APOSOR470145-1G
Pentane-2,3,4-trione 3-oxime
Pentane-2,3,4-trione 3-oxime
Formel: C₅H₇NO₃
Molecular Weight: 129.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00466577
CAS-Nummer: 29917-12-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR