5-(2-Ethoxyphenyl)-2H-pyrazol-3-ylamine hydrochloride
Lieferant: Apollo Scientific
Synonyme:
5-Amino-3-(2-ethoxyphenyl)pyrazole hydrochloride
OR470202-1G
OR470202-5G
APOSOR470202-1GEA
397
EUR
APOSOR470202-1G
APOSOR470202-5G
5-(2-Ethoxyphenyl)-2H-pyrazol-3-ylamine hydrochloride
5-(2-Ethoxyphenyl)-2H-pyrazol-3-ylamine hydrochloride
Formel:
C₁₁H₁₄ClN₃O Molecular Weight: 239.71 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11506471 CAS-Nummer: 1240134-93-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...