Ethyl-2-cyclopropyl-1,3-oxazole-4-carboxylate

Lieferant: Apollo Scientific

OR470245-5G OR470245-1G
APOSOR470245-5GEA 1210 EUR
APOSOR470245-5G APOSOR470245-1G
Ethyl-2-cyclopropyl-1,3-oxazole-4-carboxylate
Ethyl-2-cyclopropyl-1,3-oxazole-4-carboxylate
Formel: C₉H₁₁NO₃
Molecular Weight: 181,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11976754
CAS-Nummer: 1060816-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR