Ethyl-2-chloro-3-pyridylglyoxylate 95%

Lieferant: Apollo Scientific
Achtung

OR470285-1G OR470285-5G
APOSOR470285-1GEA 197 EUR
APOSOR470285-1G APOSOR470285-5G
Ethyl-2-chloro-3-pyridylglyoxylate 95%
Ethyl-2-chloro-3-pyridylglyoxylate
Formel: C₉H₈ClNO₃
Molecular Weight: 213.617 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781194
CAS-Nummer: 902837-56-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR