3-[(Cyclopropylcarbonyl)amino]-4-methylbenzoic acid

Lieferant: Apollo Scientific

OR470302-1G OR470302-5G
APOSOR470302-1GEA 245 EUR
APOSOR470302-1G APOSOR470302-5G
3-[(Cyclopropylcarbonyl)amino]-4-methylbenzoic acid
3-[(Cyclopropylcarbonyl)amino]-4-methylbenzoic acid
Formel: C₁₂H₁₃NO₃
Molecular Weight: 219.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08569890
CAS-Nummer: 915923-66-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR