[(5-Methyl-1,3-thiazol-2-yl)methyl]amine dihydrochloride

Lieferant: Apollo Scientific

Synonyme: 2-(Aminomethyl)-5-methylthiazole

OR470664-1G OR470664-250MG
APOSOR470664-1GEA 632 EUR
APOSOR470664-1G APOSOR470664-250MG
[(5-Methyl-1,3-thiazol-2-yl)methyl]amine dihydrochloride
[(5-Methyl-1,3-thiazol-2-yl)methyl]amine dihydrochloride
Formel: C₅H₁₀Cl₂N₂S
Molecular Weight: 201,12 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18071410
CAS-Nummer: 921091-08-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR