3-Methyladenin

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Methyl-3H-adenin, 6-Amino-3-methylpurin

OR470734-10MG OR470734-5G
APOSOR470734-10MGEA 21.9 EUR
APOSOR470734-10MG APOSOR470734-5G
3-Methyladenin
3-Methyladenin
Formel: C₆H₇N₅
Molecular Weight: 149,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010531
CAS-Nummer: 5142-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR