(Ethylsulphonyl)acetic acid

Lieferant: Apollo Scientific

OR470954-1G OR470954-5G
APOSOR470954-1GEA 306 EUR
APOSOR470954-1G APOSOR470954-5G
(Ethylsulphonyl)acetic acid
(Ethylsulphonyl)acetic acid
Formel: C₄H₈O₄S
Molecular Weight: 152,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09864550
CAS-Nummer: 141811-44-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR