Methyl-4-aminonicotinat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 4-aminonicotinate
OR4711-1G
OR4711-5G
APOSOR4711-1GEA
55.3
EUR
APOSOR4711-1G
APOSOR4711-5G
Methyl-4-aminonicotinat 98%
Methyl-4-aminonicotinat
Formel:
C₇H₈N₂O₂ Molecular Weight: 152.15 g/mol Schmelzpunkt: 172…174 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00955996 CAS-Nummer: 16135-36-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...