4-Chlor-2,6-dinitrobenzoesäure
Lieferant: Apollo Scientific
OR471102-1G
OR471102-5G
APOSOR471102-1GEA
275
EUR
APOSOR471102-1G
APOSOR471102-5G
4-Chlor-2,6-dinitrobenzoesäure
4-Chloro-2,6-dinitrobenzoic acid
Formel:
C₇H₃ClN₂O₆ Lagertemperatur: Raumtemperatur |
MDL:
MFCD02249880 CAS-Nummer: 95192-57-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...