3-Brom-5-formylpyridin 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromopyridine-5-carboxaldehyde, 5-Bromopyridine-3-carboxaldehyde, 5-Bromonicotinaldehyde, 5-Bromo-3-pyridinecarboxaldehyde

OR4712-25G
APOSOR4712-25GEA 81.2 EUR
APOSOR4712-25G
3-Brom-5-formylpyridin 97+%
3-Brom-5-formylpyridin
Formel: C₆H₄BrNO
Molecular Weight: 186,01 g/mol
Schmelzpunkt: 98…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03265758
CAS-Nummer: 113118-81-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR