2-Brom-5-hydroxypyridin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Brom-5-pyridinol

OR4713-5G OR4713-25G OR4713-100G
APOSOR4713-5GEA 21.9 EUR
APOSOR4713-5G APOSOR4713-25G APOSOR4713-100G
2-Brom-5-hydroxypyridin 98%
2-Brom-5-hydroxypyridin
Formel: C₅H₄BrNO
Molecular Weight: 174 g/mol
Schmelzpunkt: 133…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04114184
CAS-Nummer: 55717-45-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR