[(4-Methyl-1,3-thiazol-5-yl)methyl]amine dihydrochloride

Lieferant: Apollo Scientific

Synonyme: 5-(Aminomethyl)-4-methylthiazole

OR471301-5G OR471301-1G OR471301-250MG
APOSOR471301-5GEA 1450 EUR
APOSOR471301-5G APOSOR471301-1G APOSOR471301-250MG
[(4-Methyl-1,3-thiazol-5-yl)methyl]amine dihydrochloride
[(4-Methyl-1,3-thiazol-5-yl)methyl]amine dihydrochloride
Formel: C₅H₁₀Cl₂N₂S
Molecular Weight: 201,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18071394
CAS-Nummer: 857549-15-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR