Tetrakis(4-nitrophenyl)methane
Lieferant: Apollo Scientific
|
Achtung
|
OR471383-5G
OR471383-1G
APOSOR471383-5GEA
1220
EUR
APOSOR471383-5G
APOSOR471383-1G
Tetrakis(4-nitrophenyl)methane
Tetrakis(4-nitrophenyl)methane
Formel:
C₂₅H₁₆N₄O₈ Molecular Weight: 500,42 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12546931 CAS-Nummer: 60532-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...