2-[(4-Methylphenyl)sulphonyl]benzaldehyde

Lieferant: Apollo Scientific

Synonyme: 2-Tosylbenzaldehyde

OR471485-1G OR471485-5G
APOSOR471485-1GEA 306 EUR
APOSOR471485-1G APOSOR471485-5G
2-[(4-Methylphenyl)sulphonyl]benzaldehyde
2-[(4-Methylphenyl)sulphonyl]benzaldehyde
Formel: C₁₄H₁₂O₃S
Molecular Weight: 260,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03998747
CAS-Nummer: 171503-27-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR