Boc-Glycin-tert-Butyl-Ester 98%
Lieferant: Apollo Scientific
Synonyme:
Boc-Gly-OtBu, N-Boc-glycine tert-Butyl Ester
OR471611-100G
APOSOR471611-100GEA
75.4
EUR
APOSOR471611-100G
Boc-Glycin-tert-Butyl-Ester 98%
Boc-Glycin-tert-Butyl-Ester
Formel:
C₁₁H₂₁NO₄ Molecular Weight: 231,29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191866 CAS-Nummer: 111652-20-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...