2-Methoxy-5-methylpyridin 98%
Lieferant: Apollo Scientific
OR471639-100G
OR471639-25G
APOSOR471639-100GEA
237
EUR
APOSOR471639-100G
APOSOR471639-25G
2-Methoxy-5-methylpyridin 98%
2-Methoxy-5-methylpyridine
Formel:
C₇H₉NO Molecular Weight: 123,1500 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08703669 CAS-Nummer: 13472-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...