1-Isochromanone 98%

Lieferant: Apollo Scientific

Synonyme: 3,4-Dihydro-1H-2-benzopyran-1-one, Isochroman-1-one

OR471647-1G OR471647-25G OR471647-5G
APOSOR471647-1GEA 26.1 EUR
APOSOR471647-1G APOSOR471647-25G APOSOR471647-5G
1-Isochromanone 98%
1-Isochromanone
Formel: C₉H₈O₂
Molecular Weight: 148,16 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00799232
CAS-Nummer: 4702-34-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR