(S)-β-Phenylalaninol 95%
Lieferant: Apollo Scientific
Synonyme:
(S)-3-Amino-3-phenyl-1-propanol, (S)-3-Amino-3-phenylpropan-1-ol
OR471660-1G
OR471660-5G
APOSOR471660-1GEA
59.7
EUR
APOSOR471660-1G
APOSOR471660-5G
(S)-β-Phenylalaninol 95%
(S)-β-Phenylalaninol
Formel:
C₉H₁₃NO Molecular Weight: 151,2056 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01311768 CAS-Nummer: 82769-76-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...