5,6-Dichlor-3-pyridinol 98%
Lieferant: Apollo Scientific
Synonyme:
5,6-Dichloro-3-pyridinol, 2,3-Dichloro-5-hydroxypyridine
OR471663-5G
APOSOR471663-5GEA
190
EUR
APOSOR471663-5G
5,6-Dichlor-3-pyridinol 98%
5,6-Dichloropyridin-3-ol
Formel:
C₅H₃Cl₂NO Molecular Weight: 163,9900 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09835219 CAS-Nummer: 110860-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...