4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic acid 97%

Lieferant: Apollo Scientific

Synonyme: 4-Amino-5-chlorocoumaran-7-carboxylic acid, 4-Amino-5-chloro-2,3-dihydro-1-benzofuran-7-carboxylic acid, 4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic Acid

OR471664-5G
APOSOR471664-5GEA 101 EUR
APOSOR471664-5G
4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic acid 97%
4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic acid
Formel: C₉H₈ClNO₃
Molecular Weight: 213,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12923204
CAS-Nummer: 123654-26-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR