5-Hydroxy-2-iodbenzaldehyd 95%
Lieferant: Apollo Scientific
OR471672-1G
OR471672-5G
APOSOR471672-1GEA
169
EUR
APOSOR471672-1G
APOSOR471672-5G
5-Hydroxy-2-iodbenzaldehyd 95%
5-Hydroxy-2-iodbenzaldehyd
Formel:
C₇H₅IO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09025923 CAS-Nummer: 50765-11-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...