4-(4-Bromophenyl)pyridine 98%

Lieferant: Apollo Scientific

OR471674-5G
APOSOR471674-5GEA 87 EUR
APOSOR471674-5G
4-(4-Bromophenyl)pyridine 98%
4-(4-Bromophenyl)pyridine
Formel: C₁₁H₈BrN
Molecular Weight: 234,10 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00956748
CAS-Nummer: 39795-60-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR