2-Chloro-4-methoxynicotinic acid 95%

Lieferant: Apollo Scientific

OR471685-5G
APOSOR471685-5GEA 98.6 EUR
APOSOR471685-5G
2-Chloro-4-methoxynicotinic acid 95%
2-Chloro-4-methoxynicotinic acid
Formel: C₇H₆ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18378504
CAS-Nummer: 394729-98-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR