3-(Methoxycarbonyl)-2-methylphenylboronsäurepinakolester 97%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

OR471712-1G OR471712-5G
APOSOR471712-1GEA 114 EUR
APOSOR471712-1G APOSOR471712-5G
3-(Methoxycarbonyl)-2-methylphenylboronsäurepinakolester 97%
3-(Methoxycarbonyl)-2-methylphenylboronsäurepinakolester
Formel: C₁₅H₂₁BO₄
Molecular Weight: 276,1358 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13190786
CAS-Nummer: 955929-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR