6-Brom-2-chlor-3-pyridinamin

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-6-bromo-2-chloropyridine

OR4734-1G OR4734-5G OR4734-10G OR4734-2.5G
APOSOR4734-1GEA 21.9 EUR
APOSOR4734-1G APOSOR4734-5G APOSOR4734-10G APOSOR4734-2.5G
6-Brom-2-chlor-3-pyridinamin
6-Bromo-2-chloropyridin-3-amine
Formel: C₅H₄BrClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04971930
CAS-Nummer: 169833-70-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR