3-Amino-6-chlorpyridazin 98+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chlorpyridazin-3-amin

OR4754-100G OR4754-25G
APOSOR4754-100GEA 31.9 EUR
APOSOR4754-100G APOSOR4754-25G
3-Amino-6-chlorpyridazin 98+%
3-Amino-6-chlorpyridazin
Formel: C₄H₄ClN₃
Molecular Weight: 129,55 g/mol
Schmelzpunkt: 226…228 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051506
CAS-Nummer: 5469-69-2
EINECS: 226-796-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR