2-Methoxy-5-pyrimidinboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Methoxypyrimidin-5-yl)boronic acid

OR4771-1G OR4771-5G OR4771-25G
APOSOR4771-1GEA 21.9 EUR
APOSOR4771-1G APOSOR4771-5G APOSOR4771-25G
2-Methoxy-5-pyrimidinboronsäure
2-Methoxy-5-pyrimidineboronic acid
Formel: C₅H₇BN₂O₃
Molecular Weight: 153,9317 g/mol
Schmelzpunkt: 159…161 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03094664
CAS-Nummer: 628692-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR