Methyl-4,6-dichlorpicolinat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 4,6-dichloropyridine-2-carboxylate, Methyl 4,6-dichloropicolinate
OR47835-1G
OR47835-250MG
OR47835-5G
APOSOR47835-1GEA
63.8
EUR
APOSOR47835-1G
APOSOR47835-250MG
APOSOR47835-5G
Methyl-4,6-dichlorpicolinat
Methyl-4,6-dichlorpicolinat
Formel:
C₇H₅Cl₂NO₂ Molecular Weight: 206,0261 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12965121 CAS-Nummer: 98273-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...