4-Hydroxy-3-nitrobenzylalkohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Hydroxy-3-nitrophenyl)methanol

OR47852-25G OR47852-100G
APOSOR47852-25GEA 56.7 EUR
APOSOR47852-25G APOSOR47852-100G
4-Hydroxy-3-nitrobenzylalkohol
4-Hydroxy-3-nitrobenzylalkohol
Formel: C₇H₇NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00024249
CAS-Nummer: 41833-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR