2,5-Dichloro-4-aminophenol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Amino-2,5-dichlorophenol
OR47853-1G
OR47853-5G
APOSOR47853-1GEA
139
EUR
APOSOR47853-1G
APOSOR47853-5G
2,5-Dichloro-4-aminophenol
2,5-Dichloro-4-aminophenol
Formel:
C₆H₅Cl₂NO Molecular Weight: 178,0200 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09743861 CAS-Nummer: 50392-39-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...