1H-Indazole-7-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR48020-1G OR48020-250MG
APOSOR48020-1GEA 325 EUR
APOSOR48020-1G APOSOR48020-250MG
1H-Indazole-7-carbonitrile
1H-Indazole-7-carbonitrile
Formel: C₈H₅N₃
Schmelzpunkt: 160 °C
Lagertemperatur: Kühlschrank
MDL: MFCD10696804
CAS-Nummer: 256228-64-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR