Isoquinoline-1,3(2H,4H)-dione 95%

Lieferant: Apollo Scientific

OR480209-1G OR480209-500MG OR480209-5G
APOSOR480209-1GEA 326 EUR
APOSOR480209-1G APOSOR480209-500MG APOSOR480209-5G
Isoquinoline-1,3(2H,4H)-dione 95%
Isoquinoline-1,3(2H,4H)-dione
Formel: C₉H₇NO₂
Molecular Weight: 161.1600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234966
CAS-Nummer: 4456-77-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR