2-Amino-4-chlor-6-methylpyridin 97%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-4-chloro-6-picoline, 4-Chloro-6-methylpyridin-2-amine

OR480212-1G OR480212-250MG OR480212-5G
APOSOR480212-1GEA 37.7 EUR
APOSOR480212-1G APOSOR480212-250MG APOSOR480212-5G
2-Amino-4-chlor-6-methylpyridin 97%
2-Amino-4-chloro-6-methylpyridine
Formel: C₆H₇ClN₂
Molecular Weight: 142.5862 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00800671
CAS-Nummer: 36340-61-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR