(S)-N-3-Cyanophenylalanine 97%

Lieferant: Apollo Scientific

Synonyme: H-Phe(3-CN)-OH

OR480220-5G
APOSOR480220-5GEA 66.7 EUR
APOSOR480220-5G
(S)-N-3-Cyanophenylalanine 97%
(S)-N-3-Cyanophenylalanine
Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01860885
CAS-Nummer: 57213-48-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR