3-Amino-5-ethyl-1H-pyrazol 97%

Lieferant: Apollo Scientific

Synonyme: 3-Amino-5-ethylpyrazole

OR480223-10G OR480223-1G OR480223-5G
APOSOR480223-10GEA 207 EUR
APOSOR480223-10G APOSOR480223-1G APOSOR480223-5G
3-Amino-5-ethyl-1H-pyrazol 97%
3-Amino-5-ethyl-1H-pyrazole
Formel: C₅H₉N₃
Molecular Weight: 111,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08060982
CAS-Nummer: 1904-24-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR