2-Amino-2-(2-bromphenyl)essigsäure 95%

Lieferant: Apollo Scientific

Synonyme: H-DL-2-Phg(2-Br)-Oh

OR480224-1G OR480224-5G
APOSOR480224-1GEA 176 EUR
APOSOR480224-1G APOSOR480224-5G
2-Amino-2-(2-bromphenyl)essigsäure 95%
2-Amino-2-(2-bromphenyl)essigsäure
Formel: C₈H₈BrNO₂
Molecular Weight: 230,0586 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02662413
CAS-Nummer: 254762-66-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR