Methyl-4-amino-1-boc-piperidine-4-carboxylate 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 1-Boc-4-Aminopiperidine-4-carboxylate
OR480232-1G
OR480232-500MG
OR480232-5G
APOSOR480232-1GEA
350
EUR
APOSOR480232-1G
APOSOR480232-500MG
APOSOR480232-5G
Methyl-4-amino-1-boc-piperidine-4-carboxylate 95%
Methyl-4-amino-1-boc-piperidine-4-carboxylate
Formel:
C₁₂H₂₂N₂O₄ Molecular Weight: 258,32 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200674 CAS-Nummer: 321997-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...